Learn More
Promega Luminogenic Enzyme Substrates
The pro-luciferin substrates can be used to monitor the activity of specific isoforms of cytochrome P450 or NAT2 as indicated in the name of the substrate.
Supplier:  Promega P1621
 View more versions of this product
Catalog No. PRP1621
Description
Description
  • Luciferin-NAT2 is an excellent substrate for N-acetyltransferase 2 (NAT2), a phase II biotransformation enzyme that acetylates aromatic amine groups on xenobiotic compounds; shows little to no cross-reactivity with NAT1
  • Luciferin-MultiCYP is a promiscuous substrate that reacts with at least 21 P450 isoforms and is useful for measuring net CYP activity in a mixed population of P450s
Luciferin Detection Reagent†, ‡ must be used in combination with the above pro-luciferin substrates. Use Mfr. Nos. V8920/V8921 with P1721, P1741, P1621, P1651 and P1661. Mfr. Nos. V8930/V8931 must be used with P1671 and P1731, as these substrates are ester derivatives.
Specifications
Specifications
Monitor the activity of specific isoforms of cytochrome P450 or NAT2
Luciferin-4A Substrate
3mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.