Learn More
Promega Luminogenic Enzyme Substrates
The pro-luciferin substrates can be used to monitor the activity of specific isoforms of cytochrome P450 or NAT2 as indicated in the name of the substrate.
Specifications
For Use With (Application) | Monitor the activity of specific isoforms of cytochrome P450 or NAT2 |
---|---|
Substrate Quantity | 3mg |
Catalog Number | Mfr. No. | Substrate Quantity | Substrate Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Substrate Quantity | Substrate Type | Price | Quantity & Availability | ||||
PRP1671
|
Promega
P1671 |
3mg | Luciferin-2J2/4F12 (ester) | N/A | N/A | N/A | |||
PRP1651
|
Promega
P1651 |
3mg | Luciferin-4F2/3 Substrate | N/A | N/A | N/A | |||
PRP1661
|
Promega
P1661 |
3mg | Luciferin-4F12 Substrate | N/A | N/A | N/A | |||
PRP1741
|
Promega
P1741 |
3mg | Luciferin-3A7 Substrate | N/A | N/A | N/A | |||
PRP1621
|
Promega
P1621 |
3mg | Luciferin-4A Substrate | N/A | N/A | N/A | |||
Description
- Pro-luciferin Substrates to Monitor Activity of Cytochrome P450 Isoforms
- Used to monitor the activity of specific isoforms of cytochrome P450 or NAT2
- Choose from a wide range of substrates
- Cat.# P1621 is discontinued.
Specifications
Monitor the activity of specific isoforms of cytochrome P450 or NAT2 | |
3mg |