Learn More
Promega Luminogenic Enzyme Substrates
The pro-luciferin substrates can be used to monitor the activity of specific isoforms of cytochrome P450 or NAT2 as indicated in the name of the substrate.
Supplier:  Promega P1741
 View more versions of this product
Catalog No. PRP1741
Description
Description
  • Pro-luciferin Substrates to Monitor Activity of Cytochrome P450 Isoforms
  • Used to monitor the activity of specific isoforms of cytochrome P450 or NAT2
  • Choose from a wide range of substrates
  • Cat.# P1621 is discontinued.
Specifications
Specifications
Monitor the activity of specific isoforms of cytochrome P450 or NAT2
Luciferin-3A7 Substrate
3mg
3mg
−30 to −10°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.