Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Luminol Blend - For Forensic Detection of Blood
GREENER_CHOICESCI_ED

Catalog No. s42863 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S42863 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S42863 Supplier Innovating Science™ Supplier No. IS5040
Only null left
Add to Cart
Add to Cart

Innovating Science™ Luminol Blend - For Forensic Detection of Blood does not require a separate hydrogen peroxide catalyst.

  • To use, add 250 mL of distilled water to the dry luminol reagent
  • Spray the area of interest with the luminol solution and observe for blood evidence
  • The special luminol formulation does not require a separate hydrogen peroxide catalyst
  • Once hydrated the solution is good for up to 2 days if kept refrigerated
  • Includes: 15 g Luminol Reagent Powder

Specifications

Certifications/Compliance SDS
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Science Education
Includes 15 g Luminol Reagent Powder
Quantity 15 g
Requires Distilled water
Dimensions (L x W x H) 2 x 2 x 7 in.
Product Type Forensic Chemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.