Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Corporation Lumit™ Detection Reagent A

Catalog No. PRVB2010 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
500 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRVB2010 500 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRVB2010 Supplier Promega Corporation Supplier No. VB2010
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The Add-and-Mix Assay Format offers a simple luminescence-based detection solution with no washing steps or immobilization to plates, beads, or other surfaces. Achieve results in 30-90 minutes with a wide dynamic range.

  • Add-and-mix assay format with no washing steps required.
  • No immobilization to plates, beads, or other surfaces needed.
  • Simple luminescence-based detection.
  • 30-90 minutes total time for the assay.
  • High sensitivity and wide dynamic range.
  • Consistent and reproducible results.
  • Suitable for various research applications.
  • Compatible with high-throughput formats.
  • Reduces hands-on time and improves workflow efficiency.
  • Reliable and efficient solution for luminescence-based assays.

Specifications

Product Type Detection Reagent
Quantity 500 Assays
Species Human
Combinability Combinable
Detection Method Luminescence
Conjugate Unconjugated
Substrate Type Luminescent substrate
Research Area Immunology, Signaling, Cytokines
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.