Learn More
Description
- Measure Neonatal Fc Receptor (FcRn) binding with no wash or transfer steps.
- Simple add-mix-read protocol.
- Results in less than one hour.
- Scalable and high throughput: perform assays in 96-well or 384-well plates.
- High sensitivity and specificity.
- Consistent and reproducible results.
- Suitable for research and clinical applications.
- Streamlined workflow reduces hands-on time.
- Reliable and efficient solution for FcRn binding assays.
- Ideal for antibody characterization and stability studies.
Specifications
Specifications
| Assay | Lumit™ FcRn Binding Immunoassay |
| Detection Method | Fluorescence |
| Immunoassay Kit Format | Homogeneous Competition Assay |
| Product Type | Kit |
| Sample Type | Cell Culture |
| Specificity | High specificity |
| Sufficient For | 100 Assays |
| Target Species | Human |
| For Use With (Equipment) | Standard plate readers such as the GloMax™ Discover System and other compatible luminometers |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.