Learn More
Promega Corporation Lumit™ HMGB1 (Human/Mouse) Immunoassay
Catalog No. PRW6110 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Type:
Bioluminescent Immunoassay
Catalog No. Type
PRW6110 Bioluminescent Immunoassay
1 options
Catalog No. PRW6110 Supplier Promega Corporation Supplier No. W6110
Perform fast add-and-read assays directly on cells with the no-wash Immunoassay for HMGB1 Detection. Detects human or mouse HMGB1 using luminescent detection with just a plate-reading luminometer.
  • Perform fast add-and-read assays for HMGB1 detection directly on cells.
  • Detects human or mouse HMGB1.
  • Luminescent detection with just a plate-reading luminometer.
  • No wash steps required, simplifying the assay.
  • High sensitivity and specificity.
  • Consistent and reproducible results.
  • Suitable for various research applications.
  • Compatible with high-throughput formats.
  • Reduces hands-on time and improves workflow efficiency.
  • Ideal for cell-based assays and biomarker detection.
Specifications
Detection Method Luminescence
Type Bioluminescent Immunoassay
Cell Line U2-OS