Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Corporation Lumit™ IFN-gamma (Human) Immunoassay

Catalog No. PRW6040 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Type:
Assay
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
PRW6040 Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRW6040 Supplier Promega Corporation Supplier No. W6040
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The No-Wash Immunoassay for IFN-γ Detection offers a fast add-and-read protocol performed directly on cells. Use luminescent detection with just a plate-reading luminometer, scalable to 96- or 384-well plates.

  • Fast add-and-read protocol performed directly on cells for IFN-γ detection.
  • Luminescent detection with just a plate-reading luminometer.
  • Scalable to 96- or 384-well plates.
  • No wash steps required, simplifying the assay.
  • High sensitivity and specificity.
  • Consistent and reproducible results.
  • Suitable for various research applications.
  • Compatible with high-throughput formats.
  • Reduces hands-on time and improves workflow efficiency.
  • Ideal for cytokine detection and cell-based assays.

Specifications

Detection Method Bioluminescence
Type Assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.