Learn More
Promega Corporation Lumit™ IL-2 (Human) Immunoassay
Catalog No. PRW6021 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Type:
Bioassay
Catalog No. Type
PRW6021 Bioassay
1 options
Catalog No. PRW6021 Supplier Promega Corporation Supplier No. W6021
Add to Cart
Add to Cart
Request a Quote
Perform a fast add-and-read protocol directly on cells with the No-Wash Immunoassay for IL-2 Detection. Luminescent detection with just a plate-reading luminometer, scalable to 96- or 384-well plates.
  • Fast add-and-read protocol performed directly on cells for IL-2 detection.
  • Luminescent detection with just a plate-reading luminometer.
  • Scalable to 96- or 384-well plates.
  • No wash steps required, simplifying the assay.
  • High sensitivity and specificity.
  • Consistent and reproducible results.
  • Suitable for various research applications.
  • Compatible with high-throughput formats.
  • Reduces hands-on time and improves workflow efficiency.
  • Ideal for cytokine detection and cell-based assays.
Specifications
Detection Method Bioluminescence
Type Bioassay
Cell Line PBMC
Druggable Target Nuclear Receptors