Learn More
Promega Corporation Lumit™ Immunoassay Lysis and Detection Kit
Shop All Promega Corporation Products
Description
- Separate components for the Lumit™ Immunoassay Cellular Systems.
- Includes Lumit™ Lysis Buffer II for enhanced performance.
- Suitable for a wide range of cellular targets.
- Bundles Lumit™ Immunoassay Lysis and Detection Kit reagents with specific Lumit™ Secondary Antibody pairs.
- Cannot be used with primary antibodies from the same species.
- Sensitive, solution-based immunoassays.
- No washing steps required.
- No immobilization to plates, beads, or other surfaces needed.
- High sensitivity and specificity.
- Consistent and reproducible results.
- Streamlined workflow for ease of use.
Specifications
Specifications
| Assay | Lumit™ Immunoassay |
| Conjugate | NanoBiT™ |
| Detection Method | Fluorometric |
| Immunoassay Kit Format | Homogeneous Competition Assay |
| Product Type | Kit |
| Sample Type | Cell Culture |
| Specificity | High specificity |
| Sufficient For | 100 Assays |
| Target Species | Human |
| For Use With (Equipment) | Standard plate readers such as the GloMax™ Discover System and other compatible luminometers |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.