Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Corporation Lumit™ Immunoassay Lysis and Detection Kits and Secondary Antibodies

Catalog No. PRW1022 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
300 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRW1022 300 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRW1022 Supplier Promega Corporation Supplier No. W1022
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Enhance performance across a wide range of cellular targets with the Lumit™ Immunoassay Lysis and Detection Kits and Secondary Antibodies, sensitive, solution-based immunoassays—no washing steps, no immobilization to plates, beads or other surfaces.

  • Separate components for the Lumit™ Immunoassay Cellular Systems.
  • Includes Lumit™ Lysis Buffer II for enhanced performance.
  • Suitable for a wide range of cellular targets.
  • Bundles Lumit™ Immunoassay Lysis and Detection Kit reagents with specific Lumit™ Secondary Antibody pairs.
  • Cannot be used with primary antibodies from the same species.
  • Sensitive, solution-based immunoassays.
  • No washing steps required.
  • No immobilization to plates, beads, or other surfaces needed.
  • High sensitivity and specificity.
  • Consistent and reproducible results.

Specifications

Product Type Detection Reagent
Quantity 300 μg
Species Human
Combinability Combinable
Detection Method Luminescence
Conjugate Unconjugated
Substrate Type Luminescent substrate
Research Area Immunology, Signaling, Cytokines
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.