Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lung Cancer Marker Antibody (MOC-52) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB121610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB121610 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB121610 Supplier Novus Biologicals Supplier No. NBP311523
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lung Cancer Marker MOC-52 Monoclonal antibody specifically detects Lung Cancer Marker MOC-52 in Human samples. It is validated for Immunofluorescence, Immunofluorescence, Immunocytochemistry

Specifications

Antigen Lung Cancer Marker MOC-52
Applications Immunofluorescence, Immunocytochemistry
Classification Monoclonal
Clone MOC-52
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml, Immunocytochemistry 0.5 - 1.0 ug/ml
Formulation 10 mM PBS
Host Species Mouse
Immunogen Lung Cancer-associated antigen isolated from small cell lung carcinoma-derived cell line
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.