Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lung Cancer Marker Antibody (MOC-52), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311523FR
Description
Lung Cancer Marker Monoclonal antibody specifically detects Lung Cancer Marker in Human samples. It is validated for ImmunofluorescenceSpecifications
Lung Cancer Marker | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Lung Cancer-associated antigen isolated from small cell lung carcinoma-derived cell line | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunofluorescence | |
MOC-52 | |
Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction