Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lung Cancer Marker Antibody (MOC-52), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | Lung Cancer Marker MOC-52 |
---|---|
Clone | MOC-52 |
Concentration | 0.2 mg/mL |
Dilution | SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml, Immunocytochemistry 0.5 - 1.0 ug/ml |
Applications | SDS-Page, Immunofluorescence, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24527700
![]() |
Novus Biologicals
NBP2452770.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP24527701
![]() |
Novus Biologicals
NBP2452770.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP24527702
![]() |
Novus Biologicals
NBP2452770.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Immunocytochemistry, Immunofluorescence
Lung Cancer Marker MOC-52 Monoclonal specifically detects Lung Cancer Marker MOC-52 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunocytochemistry.Specifications
Lung Cancer Marker MOC-52 | |
0.2 mg/mL | |
SDS-Page, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Cancer, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
Lung Cancer-associated antigen isolated from small cell lung carcinoma-derived cell line. | |
Primary | |
Store at 4C. | |
145 kDa |
MOC-52 | |
SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml, Immunocytochemistry 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
100133941 | |
IgG1 κ | |
Protein A or G purified | |
This MAb reacts with a membrane-associated protein present in normal and malignant neuroendocrine tissues including small cell lung cancer (SCLC). It stains neural and a variable number of endocrine tissues and in the lung it reacts preferentially with SCLC and carcinoids. Its epitope is destroyed during formalin fixation. This antibody was categorized during the First International Workshop on Small Cell Lung Cancer Antigens held in London in April 1987. There are two major types of Lung Carcinoma: non-small cell, which accounts for 80% of all cases; and small cell, which accounts for roughly 20% of all lung cancers reported. The lung continues to be a customary place for cancer migration from tumors elsewhere in the body. Treatment depends on the specific cell type of the cancer, level of progression and status of the individual patient. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title