Learn More
GSC Go Science Crazy Lung Demonstration Model
SCI_ED
Catalog No. s28099
Encompass
Click to view available options
:
Lung demonstration model
Not a toy. Product intended for ages 13 and up. May contain latex!
Not a toy. Product intended for ages 13 and up. May contain latex!
Apparatus for classroom to demonstrate how lungs function. Partial vacuum inside the jar causes the lung shaped balloons to relax and contract.
  • Housed within partial vacuum
  • Plastic material to prevent breakage
Specifications
Description Housed within jar having partial vacuum
Product Type Lung Demonstration Model
Material Plastic
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.