Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Luperox™ A75FP, Benzoyl peroxide, 75% remainder water

Catalog No. 513474500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
500 g

Catalog No. 51-347-4500G

Supplier: Sigma Aldrich 513474500G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Luperox™ A75FP, Benzoyl peroxide, 75% remainder water

Specifications

Melting Point 103°C
Molecular Formula C14H10O4
Linear Formula (C6H5CO)2O2
Quantity 500 g
MDL Number MFCD00003071
Synonym Benzoyl peroxide moistened with water; BPO; Benzoyl peroxide; Diacyl peroxide; Dibenzoyl peroxide
Molecular Weight (g/mol) 242.23
Percent Purity 75%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.