Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LURAP1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555072
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555072 Supplier Invitrogen™ Supplier No. PA555072
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVPESLVRSL RGEEPVPRER DRDPCGGSGG GGGGGGGCSS SSSYCSFPPS LSSSSSSSPT SGSPRGSHSS ALERLETKLH LLRQEMVNLR ATDVRLMRQL LVINESIESI KW Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen LURAP1L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LURAP1L
Gene Accession No. Q8IV03
Gene Alias 1110029A09Rik; amplimer WI-13707, mouse homolog; AV077978; AV175137; bA3L8.2; bM350F23.1; C9orf150; ci150; D4Bwg0951e; HYST0841; leucine repeat adaptor protein 35b; leucine rich adaptor protein 1 like; leucine rich adaptor protein 1-like; LRAP35b; LURAP1L; RGD1308059; WI-13707
Gene Symbols LURAP1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LURAP1L. Recombinant protein control fragment (Product #RP-90371).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.