Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Luteinizing Hormone-beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102674 Supplier Invitrogen™ Supplier No. PA5102674
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LHB.

This gene is a member of the glycoprotein hormone beta chain family and encodes the beta subunit of luteinizing hormone (LH). Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. LH is expressed in the pituitary gland and promotes spermatogenesis and ovulation by stimulating the testes and ovaries to synthesize steroids. The genes for the beta chains of chorionic gonadotropin and for luteinizing hormone are contiguous on chromosome 19q13.3. Mutations in this gene are associated with hypogonadism which is characterized by infertility and pseudohermaphroditism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Luteinizing Hormone-beta
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LHB
Gene Accession No. O09108, P01229, P01230
Gene Alias aGSU; alphaGSU; alpha-GSU; alphaSU; Anterior pituitary glycoprotein hormones common subunit alpha; Cga; Cga1; CG-alpha; CGB4; Choriogonadotropin alpha chain; Chorionic gonadotrophin subunit alpha; chorionic gonadotropin, alpha polypeptide; Follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; follitropin; Follitropin alpha chain; FSHA; FSH-alpha; FSH-B; FSH-beta; Glycoprotein hormones alpha chain; glycoprotein hormones, alpha polypeptide; glycoprotein hormones, alpha subunit; GPHa; GPHA1; GPHalpha; HCG; HH23; hLHB; interstitial cell stimulating hormone, beta chain; Interstitial cell-stimulating hormone; leutenising hormone beta; leutropin; LH; LH alpha 1-1; LH alpha 1-2; LH alpha 1-3; LH alpha 3; LH beta-1; LH beta-2; LH beta-3; LHA; LHB; LH-B; LHbeta; LSH-alpha; LSH-B; LSH-beta; Luteinizing hormone (lutropin) subunit beta; Luteinizing hormone alpha chain; luteinizing hormone beta; luteinizing hormone beta polypeptide; luteinizing hormone beta subunit; Luteinizing hormone subunit beta; Lutropin alpha chain; Lutropin beta chain; Lutropin subunit beta; pituitary hormone alpha subunit; precursor polypeptide; precursor polypeptide (AA -24 to 96); Thyroid-stimulating hormone alpha chain; Thyrotropin alpha chain; TSHA; TSH-alpha
Gene Symbols LHB
Host Species Rabbit
Immunogen A synthesized peptide derived from human LHB(Accession P01229), corresponding to amino acid residues R94-L141.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16866, 25329, 3972
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.