Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CGA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583097
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583097 Supplier Invitrogen™ Supplier No. PA583097
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APDVQDCPEC TLQENPFFSQ PGAPILQCMG CCFSRAYPTP LRSKKTMLVQ KNVTSESTCC VAKSYNRVTV MGGFKVENHT ACHCSTCYYH KS Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 73%.

The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CGA
Gene Accession No. P01215
Gene Alias aGSU; alphaGSU; alpha-GSU; alphaSU; Anterior pituitary glycoprotein hormones common subunit alpha; Cga; CG-ALPHA; Choriogonadotropin alpha chain; chorionic gonadotrophin subunit alpha; chorionic gonadotropin human; chorionic gonadotropin, alpha polypeptide; follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; follitropin alpha chain; FSHA; FSH-alpha; glycoprotein hormones alpha chain; glycoprotein hormones, alpha polypeptide; glycoprotein hormones, alpha subunit; GPHa; GPHA1; GPHalpha; HCG; human chorionic gonadotropin; LHA; LSH-alpha; luteinizing hormone alpha chain; Lutropin alpha chain; thyroid-stimulating hormone alpha chain; Thyrotropin alpha chain; Tsha; TSH-alpha
Gene Symbols CGA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CGA. Recombinant protein control fragment (Product #RP-94840).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.