Learn More
Lutetium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Specifications
| Color | Silver to Gray |
| Quantity | 50 x 50 mm |
| Odor | Odorless |
| Packaging | Mylar under Argon |
| Weight | ≈0.62g/25x25mm |
| Form | Foil |
| UN Number | UN3178 |
| Percent Purity | 99.9% |
| Assay | (REO) |
| Chemical Name or Material | Lutetium foil, 0.1mm (0.004 in.) thick |
Safety and Handling
| Hazard Category | H228 |
| Hazard Statement | GHS H Statement H260-H228 In contact with water releases flammable gases which may ignite spontaneously. Flammable solid. |
| Precautionary Statement | P210-P240-P241-P280-P370+P378q |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S. |
| EINECSNumber | 231-103-0 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.