Alfa Aesar™ Lutetium(III) acetate hydrate, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1457103

Quantity 1g

 View more versions of this product

Catalog No. AA1457103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 207500-05-8
    Formula Weight 352.11 (Anhydrous)
    MDL Number MFCD00211310
    Chemical Name or Material Lutetium(III) acetate hydrate
    Grade Reacton™

    Specifications

    Formula Weight 352.11 (Anhydrous)
    Physical Form Crystalline Aggregates
    Sensitivity Hygroscopic
    MDL Number MFCD00211310
    TSCA TSCA
    CAS 207500-05-8
    Quantity 1g
    Chemical Name or Material Lutetium(III) acetate hydrate
    Assay 99.9%, (REO)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit