Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lutetium(III) oxide, 99.9% (REO), Thermo Scientific™

Catalog No. p-7050231
Click to view available options
Quantity:
1 g

Catalog No. AA3624403

Supplier: Thermo Scientific Chemicals 3624403

Only null left
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12032-20-1
Molecular Formula Lu2O3
Molecular Weight (g/mol) 397.93
MDL Number MFCD00011100
InChI Key UGBIHFMRUDAMBY-UHFFFAOYSA-N
Synonym lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
IUPAC Name dilutetium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Lu+3].[Lu+3]

Specifications

Melting Point 2487°C
Physical Form Sintered Lump
Quantity 1 g
Merck Index 14,5615
Solubility Information Insoluble in water.
Formula Weight 397.94
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 9.420 g/mL
Assay (REO)
Chemical Name or Material Lutetium(III) oxide
Show More Show Less
Hazard Category H315-H320-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-764-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.