Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-1 alpha/beta (Lutikizumab Biosimilar) Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA542121
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542121 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542121 Supplier Invitrogen™ Supplier No. MA542121
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level <0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt. Isotype of this product is: IgG1, kappa (DVD-Ig) This product is a Humanized mouse antibody.

IL-1a (Interleukin-1 alpha) is a proinflammatory cytokine produced mainly by monocytes, macrophages, and dendritic cells. Encoded by the IL1A gene on chromosome 2q13, it signals through IL-1RI and IL-1RII receptors, which are shared with IL-1B. IL-1a plays a key role in innate immunity by inducing other proinflammatory cytokines and initiating acute-phase responses. It regulates NF-kB and MAPK pathways and stimulates expression of IL-6, COX-2, and nitric oxide. IL-1a is synthesized as a proprotein that can be processed and released upon cell injury, contributing to inflammation and apoptosis. Its activity is controlled by IL-1 receptor antagonist (IL-1RA), which competitively blocks receptor binding. Elevated IL-1a levels are associated with chronic inflammatory diseases such as rheumatoid arthritis, psoriasis, and multiple sclerosis. The IL1A gene is part of an interleukin-1 family gene cluster on chromosome 2.IL-1b (Interleukin-1 beta) is another potent proinflammatory cytokine produced by similar immune cells. It is synthesized as an inactive 31 kDa precursor (pro-IL-1b) in response to inflammatory stimuli. Activation requires inflammasomes, which trigger caspase-1-mediated cleavage into the active 17 kDa form. Unlike many secreted proteins, IL-1b lacks a classical signal peptide and is released through nontraditional secretion pathways. IL-1b signals through IL-1RI and IL-1RII and is also regulated by IL-1RA. It promotes cytokine production and acute-phase responses during infection and tissue injury. Elevated IL-1b levels are linked to numerous chronic inflammatory conditions, and IL-1b-neutralizing therapies have important clinical applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1 alpha/beta (Lutikizumab Biosimilar)
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias ABT-981
Host Species Human
Immunogen Human IL1A /IL1B.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.