Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LUZP1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521935

Catalog No. PIPA521935


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, H1299, HeLa, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene includes two alternatively spliced transcript variants, which encode different isoforms. The function of this gene has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LUZP1
Polyclonal
Unconjugated
LUZP1
2700072H04Rik; AI266952; Leucine zipper motif-containing protein; leucine zipper protein 1; Luzp; LUZP1; mFLJ00226; RP1-184J9.3
Rabbit
Antigen affinity chromatography
RUO
7798
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.28 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q86V48
LUZP1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human LUZP1. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.