Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LUZP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555894 Supplier Invitrogen™ Supplier No. PA555894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLLASQLEEI KLQIKKQKEL ENGEVEGEDA FLSSKGRHER TKFRGHGSEA SVSKHTAREL SPQHKRERLR NREFALNNEN YSLSNRQVSS PSFT Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 77%.

This gene includes two alternatively spliced transcript variants, which encode different isoforms. The function of this gene has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LUZP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LUZP1
Gene Accession No. Q86V48
Gene Alias 2700072H04Rik; AI266952; Leucine zipper motif-containing protein; leucine zipper protein 1; Luzp; LUZP1; mFLJ00226; RP1-184J9.3
Gene Symbols LUZP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LUZP1. Recombinant protein control fragment (Product #RP-94448).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7798
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.