Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LUZP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | LUZP2 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
LUZP2 Polyclonal specifically detects LUZP2 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
LUZP2 | |
Polyclonal | |
Rabbit | |
Human | |
338645 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DYEELEKQLKEVFKERSTILRQLTKTSRELDGIKVNLQSLKNDEQSAKTDVQKLLELGQKQREEMKSLQEALQNQLKETSEKAEKHQATINFLKTEVERKSKMIRDL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
KFSP2566, LUZP2 leucine zipper protein 2, PRO6246 | |
LUZP2 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title