Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LUZP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561168
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561168 Supplier Invitrogen™ Supplier No. PA561168
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DYEELEKQLK EVFKERSTIL RQLTKTSREL DGIKVNLQSL KNDEQSAKTD VQKLLELGQK QREEMKSLQE ALQNQLKETS EKAEKHQATI NFLKTEVERK SKMIRDL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

This gene encodes a leucine zipper protein. This protein is deleted in some patients with Wilms tumor-Aniridia-Genitourinary anomalies-mental Retardation (WAGR) syndrome. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Oct 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LUZP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LUZP2
Gene Accession No. Q86TE4
Gene Alias KFSP2566; Leucine zipper protein 2; LUZP2; PRO6246; UNQ2566/PRO6246
Gene Symbols LUZP2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human LUZP2 (Product #RP-97531)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338645
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.