Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LUZP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565223
Description
Immunogen sequence: DYEELEKQLKE VFKERSTILR QLTKTSRELD GIKVNLQSLK NDEQSAKTDV QKLLELGQKQ REEMKSLQEA LQNQLKETSE KAEKHQATIN FLKTEVERKS KMIRDL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
This gene encodes a leucine zipper protein. This protein is deleted in some patients with Wilms tumor-Aniridia-Genitourinary anomalies-mental Retardation (WAGR) syndrome. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Oct 2011].
Specifications
LUZP2 | |
Polyclonal | |
Unconjugated | |
LUZP2 | |
KFSP2566; Leucine zipper protein 2; LUZP2; PRO6246; UNQ2566/PRO6246 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
338645 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86TE4 | |
LUZP2 | |
Recombinant Human LUZP2. Recombinant protein control fragment (Product #RP-97531). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction