Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LUZP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562528
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562528 Supplier Invitrogen™ Supplier No. PA562528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQEKCSDNYE AQAEKNQGQS EGNQHQSEGN PDKSEESQGQ PEENHHSERS RNHLERSLSQ SDRSQGQLKR HHPQYERSHG QYKRSHGQSE RSHGHS Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 28%.

Export adapter involved in mRNA nuclear export in cancer cells. Binds and enhances the RNA-binding activity of the nuclear RNA export factor NXF1. Can restore mRNA export function in cells compromised by loss of mRNA export adapters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LUZP4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LUZP4
Gene Accession No. Q9P127
Gene Alias Cancer/testis antigen 28; CT28; CT-28; CT-8; HOM-TES-85; HOM-TES-85 tumor antigen; leucine zipper protein 4; LUZP4; Tumor antigen HOM-TES-85
Gene Symbols LUZP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LUZP4. Recombinant protein control fragment (Product #RP-100049).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51213
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.