Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LUZP6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567481
Description
Immunogen sequence: VISYALYQVQT GSLPVYSSVL TKSPLQLQTV IYRLIVQIQH LNIPSSSSTH SS Highest antigen sequence identity to the following orthologs - mouse 30%, rat 28%.
A bi-cistronic transcript encodes the products of both the myotrophin and leucine zipper protein 6 genes, which are located on chromosome 7. A cryptic ORF at the 3' end of the myotrophin transcript uses a novel internal ribosome entry site and a non-AUG translation initiation codon to produce leucine zipper protein 6, a 6.4 kDa tumor antigen that is associated with myeloproliferative disease.
Specifications
LUZP6 | |
Polyclonal | |
Unconjugated | |
LUZP6 | |
leucine zipper protein 6; LUZP6; MPD6; MTPNUT; Myeloproliferative disease-associated 6 kDa antigen; myeloproliferative disease-associated antigen, 6-kD; myeloproliferative disease-associated SEREX antigen; myotrophin 3'UTR transcript | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
767558 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q538Z0 | |
LUZP6 | |
Recombinant Human LUZP6. Recombinant protein control fragment (Product #RP-108134). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction