Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LXR alpha + LXR beta Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501984218
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4218 Supplier Bioss Supplier No. BS18451R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NR1H3 (oxysterols receptor LXR-alpha) is a nuclear receptor that exhibits a ligand-dependent transcriptional activiation activity. Its interaction with retinoic acid receptor (RXR) shifts RXR from its role as a silent DNA-binding partner to an active ligand-binding subunit in mediating retinoid responses through target genes defined by LXRES. LXRES are DR4-type response elements characterized by direct repeats of two similar hexanucleotide half-sites spaced by four nucleotides. NR1H3 plays an important role in the regulation of cholesterol homeostasis, regulating cholesterol uptake through MYLIP-dependent ubiqutination of LDLR, VLDLR, and LRP8. It also interplays functionally with RORA for the regulation of genes involves in liver metabolism.

Specifications

Antigen LXR alpha + LXR beta
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NR1H2
Gene Accession No. P55055, Q13133, Q60644, Q62685, Q62755, Q9Z0Y9
Gene Alias AI194859; AU018371; liver X nuclear receptor alpha variant 1; liver X nuclear receptor beta; Liver X Receptor; Liver X receptor alpha; Liver X receptor beta; LX receptor beta; LXR; LXR alpha; LXR beta; LXRA; LXR-a; LXRalpha; LXRB; LXR-b; LXRbeta; NER; NER1; NER-I; NR1H2; Nr1h3; nuclear orphan receptor LXR-alpha; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 2; nuclear receptor subfamily 1, group H, member 3; OR-1; Orphan nuclear receptor OR-1; Oxysterols receptor LXR-alpha; Oxysterols receptor LXR-beta; Retinoid X receptor-interacting protein No.15; Rip15; RLD1; RLD-1; steroid hormone-nuclear receptor NER; ubiquitous receptor; Ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 1; ubiquitously-expressed nuclear receptor 2; Unr; Unr1; Unr2; UR
Gene Symbols NR1H2, NR1H3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LXR alpha + LXR beta.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10062, 22259, 22260, 58851, 58852, 7376
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.