Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LXR alpha + LXR beta Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS10311R

Description
NR1H3 (oxysterols receptor LXR-alpha) is a nuclear receptor that exhibits a ligand-dependent transcriptional activiation activity. Its interaction with retinoic acid receptor (RXR) shifts RXR from its role as a silent DNA-binding partner to an active ligand-binding subunit in mediating retinoid responses through target genes defined by LXRES. LXRES are DR4-type response elements characterized by direct repeats of two similar hexanucleotide half-sites spaced by four nucleotides. NR1H3 plays an important role in the regulation of cholesterol homeostasis, regulating cholesterol uptake through MYLIP-dependent ubiqutination of LDLR, VLDLR, and LRP8. It also interplays functionally with RORA for the regulation of genes involves in liver metabolism.Specifications
LXR alpha + LXR beta | |
Polyclonal | |
Unconjugated | |
NR1H2 | |
AI194859; AU018371; liver X nuclear receptor alpha variant 1; liver X nuclear receptor beta; Liver X Receptor; Liver X receptor alpha; Liver X receptor beta; LX receptor beta; LXR; LXR alpha; LXR beta; LXRA; LXR-a; LXRalpha; LXRB; LXR-b; LXRbeta; NER; NER1; NER-I; NR1H2; Nr1h3; nuclear orphan receptor LXR-alpha; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 2; nuclear receptor subfamily 1, group H, member 3; OR-1; Orphan nuclear receptor OR-1; Oxysterols receptor LXR-alpha; Oxysterols receptor LXR-beta; Retinoid X receptor-interacting protein No.15; Rip15; RLD1; RLD-1; steroid hormone-nuclear receptor NER; ubiquitous receptor; Ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 1; ubiquitously-expressed nuclear receptor 2; Unr; Unr1; Unr2; UR | |
Rabbit | |
Protein A | |
RUO | |
10062, 22259, 22260, 58851, 58852, 7376 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P55055, Q13133, Q60644, Q62685, Q62755, Q9Z0Y9 | |
NR1H2, NR1H3 | |
KLH conjugated synthetic peptide derived from human LXR alpha. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction