Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LXR alpha/NR1H3 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400157H
Description
LXR alpha/NR1H3 Polyclonal specifically detects LXR alpha/NR1H3 in Human, Mouse, Porcine, Primate samples. It is validated for Western Blot, Simple Western.Specifications
LXR | |
Polyclonal | |
Western Blot, Simple Western | |
Q13133 | |
NR1H3 | |
A synthetic peptide made to an internal portion of the human LXR protein sequence (between residues 50-150). [UniProt# Q13133] | |
0.1 mL | |
Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Signal Transduction, Transcription Factors and Regulators | |
10062 | |
Human, Mouse, Pig, Primate | |
IgG |
Western Blot | |
HRP | |
PBS with No Preservative | |
alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, porcine and monkey. Immunogen sequence has 92% identity to rat and 90% identity to chicken. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction