Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LXR alpha/NR1H3 Antibody (OTI1A5), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272550V
Description
LXR alpha/NR1H3 Monoclonal specifically detects LXR alpha/NR1H3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LXR alpha/NR1H3 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NR1H3 | |
Recombinant protein fragment corresponding to amino acids 69-329 of human LXR alpha/NR1H3 (NP_005684) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI1A5 | |
Western Blot, Immunohistochemistry | |
alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1 | |
Mouse | |
Affinity Purified | |
Diabetes Research, Lipid and Metabolism | |
10062.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction