Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LXR beta Monoclonal Antibody (K8917)
GREENER_CHOICE

Catalog No. 418400
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-840-0 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-840-0 Supplier Invitrogen™ Supplier No. 418400
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody specifically recognizes human LXR beta but does not recognize human LXR alpha. This antibody does not cross react with rat LXR beta. Not yet tested in other species.

The liver X receptors, LXRA and LXRB, form a subfamily of the nuclear receptor superfamily and are key regulators of macrophage function, controlling transcriptional programs involved in lipid homeostasis and inflammation. The inducible LXRA is highly expressed in liver, adrenal gland, intestine, adipose tissue, macrophages, lung, and kidney, whereas LXRB is ubiquitously expressed. Ligand-activated LXRs form obligate heterodimers with retinoid X receptors and regulate expression of target genes containing LXR response elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LXR beta
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K8917
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Physiological saline with 0.1% sodium azide
Gene NR1H2
Gene Accession No. P55055
Gene Alias AI194859; liver X nuclear receptor beta; liver X receptor beta; LX receptor beta; LXR; LXR beta; LXRB; LXR-b; LXRbeta; NER; NER1; NER-I; Nr1h2; NR1H2 protein; nuclear orphan receptor LXR-beta; nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; OR-1; Orphan nuclear receptor OR-1; oxysterols receptor LXR-beta; Retinoid X receptor-interacting protein No.15; RIP15; steroid hormone-nuclear receptor NER; ubiquitous receptor; ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 2; UNR; Unr2; UR
Gene Symbols NR1H2
Host Species Mouse
Immunogen Produced in BALB/c mouse ascites after inoculation with hybridoma of mouse myeloma cells (NS-1) and spleen cells derived from a BALB/c mouse immunized with Baculovirus-expressed recombinant human LXR-beta (2-86 aa).
Purification Method Ammonium sulfate precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7376
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.