Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LXR beta/NR1H2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | LXR beta/NR1H2 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
LXR beta/NR1H2 Polyclonal specifically detects LXR beta/NR1H2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LXR beta/NR1H2 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Liver X receptor beta, LXR-b, NER-I, NERliver X receptor-beta, nuclear orphan receptor LXR-beta, Nuclear receptor NER, Nuclear receptor subfamily 1 group H member 2, nuclear receptor subfamily 1, group H, member 2, oxysterols receptor LXR-beta, RIP15, steroid hormone-nuclear receptor NER, Ubiquitously-expressed nuclear receptorLXRB, UNRLX receptor beta | |
NR1H2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
P55055 | |
7376 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PDVPGTDEASSACSTDWVIPDPEEEPERKR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title