Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LXR beta Polyclonal Antibody

Catalog No. PIPA529771
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529771 Supplier Invitrogen™ Supplier No. PA529771

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, mouse brain. Predicted reactivity: Mouse (93%), Rat (92%), Rabbit (93%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The liver X receptors, LXRA and LXRB, form a subfamily of the nuclear receptor superfamily and are key regulators of macrophage function, controlling transcriptional programs involved in lipid homeostasis and inflammation. The inducible LXRA is highly expressed in liver, adrenal gland, intestine, adipose tissue, macrophages, lung, and kidney, whereas LXRB is ubiquitously expressed. Ligand-activated LXRs form obligate heterodimers with retinoid X receptors and regulate expression of target genes containing LXR response elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LXR beta
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene NR1H2
Gene Accession No. P55055, Q60644
Gene Alias AI194859; liver X nuclear receptor beta; liver X receptor beta; LX receptor beta; LXR; LXR beta; LXRB; LXR-b; LXRbeta; NER; NER1; NER-I; Nr1h2; NR1H2 protein; nuclear orphan receptor LXR-beta; nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; OR-1; Orphan nuclear receptor OR-1; oxysterols receptor LXR-beta; Retinoid X receptor-interacting protein No.15; RIP15; steroid hormone-nuclear receptor NER; ubiquitous receptor; ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 2; UNR; Unr2; UR
Gene Symbols NR1H2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human LXR beta. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22260, 7376
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.