Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LXR beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520886
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520886 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520886 Supplier Invitrogen™ Supplier No. PA520886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human lung tissue lysate. PA5-20886 can be used with blocking peptide PEP-1000.

The liver X receptors, LXRA and LXRB, form a subfamily of the nuclear receptor superfamily and are key regulators of macrophage function, controlling transcriptional programs involved in lipid homeostasis and inflammation. The inducible LXRA is highly expressed in liver, adrenal gland, intestine, adipose tissue, macrophages, lung, and kidney, whereas LXRB is ubiquitously expressed. Ligand-activated LXRs form obligate heterodimers with retinoid X receptors and regulate expression of target genes containing LXR response elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LXR beta
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NR1H2
Gene Accession No. P55055, Q60644, Q62755
Gene Alias AI194859; liver X nuclear receptor beta; liver X receptor beta; LX receptor beta; LXR; LXR beta; LXRB; LXR-b; LXRbeta; NER; NER1; NER-I; Nr1h2; NR1H2 protein; nuclear orphan receptor LXR-beta; nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; OR-1; Orphan nuclear receptor OR-1; oxysterols receptor LXR-beta; Retinoid X receptor-interacting protein No.15; RIP15; steroid hormone-nuclear receptor NER; ubiquitous receptor; ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 2; UNR; Unr2; UR
Gene Symbols NR1H2
Host Species Rabbit
Immunogen A 14 amino acid peptide from near the amino terminus human LXR-B.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22260, 58851, 7376
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.