Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LXR beta Recombinant Rabbit Monoclonal Antibody (PSH0-56)
GREENER_CHOICE

Catalog No. PIMA547617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547617 Supplier Invitrogen™ Supplier No. MA547617
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Sequence Similarities: 83% Mouse/Rat Tissue Specificity: Ubiquitous Positive control: SW480 cell lysate, HepG2 cell lysate, PC-3M cell lysate, A549 cell lysate, HeLa cell lysate, HT-29 cell lysate, HCT116 cell lysate, HEK-293 cell lysate, mouse brain tissue lysate, mouse liver tissue lysate, rat liver tissue lysate, C2C12, HCT116. Subcellular location: Nucleus.

The liver X receptors, LXRA and LXRB, form a subfamily of the nuclear receptor superfamily and are key regulators of macrophage function, controlling transcriptional programs involved in lipid homeostasis and inflammation. The inducible LXRA is highly expressed in liver, adrenal gland, intestine, adipose tissue, macrophages, lung, and kidney, whereas LXRB is ubiquitously expressed. Ligand-activated LXRs form obligate heterodimers with retinoid X receptors and regulate expression of target genes containing LXR response elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LXR beta
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone PSH0-56
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene NR1H2
Gene Accession No. P55055, Q60644, Q62755
Gene Alias AI194859; liver X nuclear receptor beta; liver X receptor beta; LX receptor beta; LXR; LXR beta; LXRB; LXR-b; LXRbeta; NER; NER1; NER-I; Nr1h2; NR1H2 protein; nuclear orphan receptor LXR-beta; nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; OR-1; Orphan nuclear receptor OR-1; oxysterols receptor LXR-beta; Retinoid X receptor-interacting protein No.15; RIP15; steroid hormone-nuclear receptor NER; ubiquitous receptor; ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 2; UNR; Unr2; UR
Gene Symbols NR1H2
Host Species Rabbit
Immunogen Synthetic peptide within human lxr beta aa 50-100/460.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22260, 58851, 7376
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.