Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-108 Monoclonal Antibody (eBio13G3-19D (13G3-19D)), NovaFluor™ Yellow 700, eBioscience™, Invitrogen™

Catalog No. M050T03Y06A
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M050T03Y06A 100 μg
M050T02Y06A 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M050T03Y06A Supplier Invitrogen™ Supplier No. M050T03Y06A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ly108, the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6), is expressed on NK cells, B cells, and T cells. Engagement of the Ly108 receptor enhances NK cell activity and increases the expression of IFN-gamma. This receptor plays a role in the inability of NK cells to kill Epstein Barr virus-infected B cells in X-linked lymphoproliferative disease. In Ly108 knock-out mice, CD4+ T cells show a diminished ability to produce IL-4 and exhibit defective neutrophil functions, indicating the importance of Ly108 in immune regulation and cellular function. Overall, Ly108 is crucial for modulating immune responses and maintaining effective immune cell activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-108
Applications Flow Cytometry
Classification Monoclonal
Clone eBio13G3-19D (13G3-19D)
Concentration 0.1 mg/mL
Conjugate NovaFluor Yellow 700
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene SLAMF6
Gene Accession No. Q9ET39
Gene Alias Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080
Gene Symbols SLAMF6
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30925
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.