Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-108 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA562389

Catalog No. PIPA562389


Only null left
Add to Cart

Description

Description

Immunogen sequence: LPLEFPAGEK VNFITWLFNE TSLAFIVPHE TKSPEIHVTN PKQGKRLNFT QSYSLQLSNL KMEDTGSYRA QISTKT Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 50%.

Ly108 is the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6). Mouse Ly108 is expressed on NK, B cells, and T cells. Activation of Ly108 receptor enhances NK cell activity and IFN-gamma expression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ly-108
Polyclonal
Unconjugated
SLAMF6
Activating NK receptor; CD352; KAL1; KAL1b; KALI; KALIb; Ly108; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTBA; NTB-A; NTBA receptor; RGD1561848; SF2000; SLAM family member 6; Slamf6; UNQ6123/PRO20080
Rabbit
Antigen Affinity Chromatography
RUO
114836
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96DU3
SLAMF6
Recombinant protein corresponding to Human Ly-108. Recombinant protein control fragment (Product #RP-101416).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.