Learn More
Tocris Bioscience™ LY 2033298

Description
α indicates the degree of allosteric enhancement when both orthosteric and allosteric sites are occupied (derived from the allosteric ternary complex model).
Specifications
Specifications
| Chemical Name or Material | 3-Amino-5-chloro-N-cyclopropyl-6-methoxy-4-methylthieno[2,3-b]pyridine-2-carboxamide |
| CAS | 886047-13-8 |
| Quantity | 25 mg |
| Molecular Formula | C13H14ClN3O2S |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.