Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 2033298
GREENER_CHOICESDP

Catalog No. 70275 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-275 5 mg
70-272-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 70-275 Supplier Tocris Bioscience™ Supplier No. 7027/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective positive allosteric modulator of M4 receptors.

LY 2033298 is a selective positive allosteric modulator of M4 receptor (KB = 200 nM, α = 35); increases the potency of acetylcholine by 40-fold. Exhibits no effect at hM1/3/5 receptors and a small effect at hM2 (KB = 1 μM α = 3.7). Potentiates other full and partial orthosteric agonists, including Carbachol, Oxotremorine-M, and McN-A343 . Does not affect binding of antagonist NMS. Active in vivo. Antipsychotic.

α indicates the degree of allosteric enhancement when both orthosteric and allosteric sites are occupied (derived from the allosteric ternary complex model).

Specifications

Chemical Name or Material 3-Amino-5-chloro-N-cyclopropyl-6-methoxy-4-methylthieno[2,3-b]pyridine-2-carboxamide
CAS 886047-13-8
Quantity 5 mg
Molecular Formula C13H14ClN3O2S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.