Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 2087101
GREENER_CHOICE

Catalog No. p-4816749
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-411-0 10 mg
41-415-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-411-0 Supplier Tocris Bioscience™ Supplier No. 4141/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potentiator of α7 nAChRs

Allosteric potentiator of α7, α4β2 and α4β4 nAChRs; displays selectivity against α3β4 nAChRs. Thought to potentiate agonist-evoked α7 responses by binding within the nAChR transmembrane region.

Chemical Identifiers

CAS 913186-74-0
Molecular Formula C15H11FN2OS2

Specifications

Quantity 10 mg
Formula Weight 318.39
Purity >98%
Molecular Weight (g/mol) 318.384
Chemical Name or Material LY 2087101
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.