Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 2886721 Hydrochloride
GREENER_CHOICE

Catalog No. 708110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-811-0 10 mg
70-815-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 70-811-0 Supplier Tocris Bioscience™ Supplier No. 7081/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective β-secretase (BACE) inhibitor

LY 2886721 Hydrochloride is a potent and selective β-secretase (BACE) inhibitor (IC50 values are 10.2 and 20.3 nM for human BACE2 and BACE1, respectively). Displays >5,000-fold selectivity for BACE over other proteases including cathepsin D, pepsin and renin. Inhibits Aβ1-40 and Aβ1-42 production in cells expressing mutated APP. Reduces hippocampal and cortical Aβ and sAPPβ levels in an Alzheimer's disease mouse model.

Specifications

Chemical Name or Material N-[3-[(4 AS,7 AS)-2-Amino-4 A,5-dihydro-4 H-furo[3,4-d][1,3 ]thiazin-7 A(7 H)-yl]-4-fluorophenyl]-5-fluoro-2-pyridinecarboxamide hydrochloride
CAS 1262036-49-6
Quantity 10 mg
Target Additional Synthetic Protease Inhibitors
Molecular Formula C18H16F2N4O2S.HCl
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.