Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals LY 294002, 98%

Catalog No. AAJ67289MA
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67289MA 10 mg
AAJ67289LB0 5 mg
AAJ67289MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ67289MA Supplier Thermo Scientific Chemicals Supplier No. J67289MA

CAS: 154447-36-6 | C19H17NO3, C19H17NO3 | 307.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154447-36-6
Molecular Formula C19H17NO3
Synonym 2-(4-morpholinyl)-8-phenyl-4H-1-benzopyran-4-one;

Specifications

Quantity 10 mg
Description White to pale yellow
Content And Storage -30°C to -10°C
Formula Weight 307.35g/mol
Form Solid
Assay 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.