Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY 320135, Tocris Bioscience™

Catalog No. p-4784122
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective CB1 inverse agonist

Potent CB1 receptor antagonist/inverse agonist (Ki = 141 nM) with greater than 70-fold selectivity over CB2 receptors (Ki > 10 μM). Structurally dissimilar from SR 141716A and AM 251. Shows weak binding to both 5-HT2 (Ki = 6.4 μM) and muscarinic receptors (Ki = 2.1 μM).

Chemical Identifiers

CAS 176977-56-3
Molecular Formula C24H17NO4
Molecular Weight (g/mol) 383.40
MDL Number MFCD00945878
InChI Key RYNSGDFWBJWWSZ-UHFFFAOYSA-N
Synonym unii-cjy03984ct, 6-methoxy-2-4-methoxyphenyl benzo b furan-3-yl 4-cyanophenyl methanone, 4-6-methoxy-2-4-methoxyphenyl benzofuran-3-carbonyl benzonitrile, 4-6-methoxy-2-4-methoxyphenyl-3-benzofuranyl carbonyl benzonitrile, 4-6-methoxy-2-4-methoxyphenyl-1-benzofuran-3-carbonyl benzonitrile, d0zu9z, gtpl742, benzonitrile, 4-6-methoxy-2-4-methoxyphenyl-3-benzofuranyl carbonyl, ly hplc
PubChem CID 5311257
IUPAC Name 4-[6-methoxy-2-(4-methoxyphenyl)-1-benzofuran-3-carbonyl]benzonitrile
SMILES COC1=CC=C(C=C1)C1=C(C(=O)C2=CC=C(C=C2)C#N)C2=CC=C(OC)C=C2O1

Specifications

Quantity 10 mg
Formula Weight 383.4
Percent Purity >98%
Chemical Name or Material LY 320135
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.