Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 341495 disodium salt
SDP

Catalog No. 40621 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-621 1 mg
40-6210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-621 Supplier Tocris Bioscience™ Supplier No. 4062/1

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective group II mGlu antagonist; disodium salt of LY 341495 (Cat. No. 1209)

Highly potent and selective group II metabotropic glutamate receptor antagonist. Disodium salt of LY 341495 (Cat. No. 1209).

Chemical Identifiers

Molecular Formula C20H17NNa2O5

Specifications

Quantity 1 mg
Formula Weight 397.33
Purity >97%
Chemical Name or Material LY 341495 disodium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.