Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ LY 367385
GREENER_CHOICESDP

Catalog No. 123750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-375-0 50 mg
12-371-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-375-0 Supplier R&D Systems™ Supplier No. 1237/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective mGlu1a antagonist

A selective mGlu1a receptor antagonist, with an IC50 value of 8.8 μM for blockade of quisqualate-induced phosphoinositide hydrolysis vs. > 100 μM for mGlu5a, and negligible action on group II and III receptors.

Chemical Identifiers

CAS 198419-91-9
Molecular Formula C10H11NO4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 209.2
Purity >99%
Molecular Weight (g/mol) 209.201
Chemical Name or Material LY 367385
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.