Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LY 404187
GREENER_CHOICESDP

Catalog No. 529750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-975-0 50 mg
52-971-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-975-0 Supplier Tocris Bioscience™ Supplier No. 5297/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective AMPA receptor positive allosteric modulator

LY 404187 is a selective AMPA receptor positive allosteric modulator (IC50 values are 0.15, 0.21, 1.66 and 5.65 μM for GluA2i, GluA4i, GluA3i and GluA1i respectively). Potentiates glutamate and AMPA-evoked currents in pre frontal cortex (PFC) pyramidal neurons in vitro. Has no effect on NMDA- or kainate-evoked currents; also has no effect on K+ or Na+ ion channels. Potentiates PFC glutamatergic synaptic transmission in vitro and in vivo. Induces neurite growth in combination with (S)-AMPA (Cat. No. 0254) in SH-SY5Y human neuroblastoma cells in vitro. Also reduces MPTP-induced toxicity in mice. Brain penetrant.

Specifications

Chemical Name or Material N-[2-(4′-Cyano[1,1′-biphenyl]-4-yl)propyl]-2-propanesulfonamide
CAS 211311-95-4
Quantity 50 mg
Target AMPA Receptor Modulators
Molecular Formula C19H22N2O2S
Formula Weight 342.46
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.